missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dianilinoethane, 97%

Product Code. 11466296
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11466296

Brand: Thermo Scientific Alfa Aesar B23098.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-61-8 | C14H16N2 | 212.30 g/mol

1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.

Solubility
Sparingly soluble in water 0.072 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-61-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
MDL Number MFCD00003019
InChI Key NOUUUQMKVOUUNR-UHFFFAOYSA-N
Synonym 1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin
PubChem CID 67422
IUPAC Name N,N'-diphenylethane-1,2-diamine
SMILES C(CNC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 64°C to 68°C
Boiling Point 228°C (12 mmHg)
Quantity 5 g
Beilstein 646740
Merck Index 14,2990
Solubility Information Sparingly soluble in water 0.072g/L @ 25°C.
Formula Weight 212.3
Percent Purity ≥98%
Chemical Name or Material 1,2-Dianilinoethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.