missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diaminopropane-N,N,N',N'-tetraacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid |
| CAS | 4408-81-5 |
| Molecular Formula | C11 H18 N2 O8 |
| Quantity | 1 g |
| Synonym | Acetic acid, (propylenedinitrilo)tetra- (6CI,7CI,8CI), N,N'-(1-Methyl-1,2-ethanediyl)bis[N-(carboxymethyl)glycine], (±)-1,2-Diaminopropanetetraacetic acid, 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid, 1,2-Diaminopropanetetraacetate, 1,2-Diaminopropanetetraacetic acid, 1,2-Propanediaminetetraacetic acid, 1,2-Propylenediamine-N,N,N',N'-tetraacetic acid, 1,2-Propylenediaminetetraacetic acid, 2,2',2'',2'''-(1,2-Propanediyldinitrilo)tetraacetic acid, DL-Propylenediaminetetraacetic acid, Dissolvine P, Dissolvine PDZ, MeEDTA, Methyl-EDTA, Propylene-1,2-diaminetetraacetic acid, Trilon FS |
| InChI Formula | InChI=1S/C11H18N2O8/c1-7(13(5-10(18)19)6-11(20)21)2-12(3-8(14)15)4-9(16)17/h7H,2-6H2,1H3,(H,14,15)(H,16,17)(H,18,19)(H,20,21) |
| SMILES | CC(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
| IUPAC Name | 2-[2-[bis(carboxymethyl)amino]propyl-(carboxymethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 306.27 |
| Formula Weight | 306.1063 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?