missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminopropane, 99%

Product Code. 15483267
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483267 1000 mL 1000mL
15453267 100 mL 100mL
15463267 250 mL 250mL
15473267 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15483267 Supplier Thermo Scientific Alfa Aesar Supplier No. B24489.AU
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-90-0 | C3H10N2 | 74.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008089
InChI Key AOHJOMMDDJHIJH-UHFFFAOYNA-N
Synonym 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine
PubChem CID 6567
ChEBI CHEBI:30630
IUPAC Name propane-1,2-diamine
SMILES CC(N)CN

Specifications

Melting Point ∼-37°C
Density 0.869
Boiling Point 119°C to 120°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.446
Quantity 1000 mL
UN Number UN2258
Beilstein 605274
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,7852
Formula Weight 74.13
Percent Purity 99%
Chemical Name or Material 1,2-Diaminopropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.