missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Deoxy-beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione (Thymidine), Mikromol™

Product Code. 17968544
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968544 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17968544 Supplier Mikromol Supplier No. MM0534.030025
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(2-Deoxy-beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione (Thymidine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thymidin
CAS 50-89-5
Analyte or Component Names Thymidin
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2 O5
Certifications/Compliance ISO 9001
Synonym Stavudine Imp. C (EP), Zidovudine USP Related Compound D, Zidovudine USP RC D, Thymidine, 1-(2-Deoxy-beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione, Zidovudine Imp. E (Pharmeuropa), 1-(2-Deoxy-beta-D-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione, 2,4(1H,3H)-Pyrimidinedione, 1-(2-deoxy-beta-D-erythro-pentofuranosyl)-5-methyl-, 2'-Deoxythymidine, 5-Methyl-2'-deoxyuridine, 5-Methyldeoxyuridine, DThyd, Deoxyribothymidine, Deoxythymidine, NSC 21548, Thymidin, Thymine 2-desoxyriboside, Thymine deoxyriboside, Uridine, 2'-deoxy-5-methyl-, dT, beta-D-Ribofuranoside, thymine-1 2-deoxy-, Thymidine, Zidovudine Related Compound D, Stavudine Impurity C, Zidovudine Impurity E
InChI Formula InChI=1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1
SMILES CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
Molecular Weight (g/mol) 242.23
Quantity 25 mg
Formula Weight 242.0903 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.