missing translation for 'onlineSavingsMsg'
Learn More

1',2'-Dehydro Dicyclomine (>90%), TRC

Product Code. 30011980
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011980 250 mg 250mg
30004787 50 mg 50mg
30011708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011980 Supplier Toronto Research Chemicals Supplier No. TRCD229530250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109158-77-2
Molecular Formula C19H33NO2
Molecular Weight (g/mol) 307.47
Synonym Cyclohexanecarboxylic acid, 1-(1-cyclohexen-1-yl)-, 2-(diethylamino)ethyl ester, Cyclohexanecarboxylic acid, 1-(1-cyclohexen-1-yl)-, 2-diethylaminoethyl ester (6CI), 2-(Diethylamino)ethyl 1,1'-bi(cyclohexan)-1′-ene-1-carboxylate
IUPAC Name 2-(diethylamino)ethyl 1-(cyclohexen-1-yl)cyclohexanecarboxylate
SMILES CCN(CC)CCOC(=O)C1(CCCCC1)C2=CCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H33NO2/c1-3-20(4-2)15-16-22-18(21)19(13-9-6-10-14-19)17-11-7-5-8-12-17/h11H,3-10,12-16H2,1-2H3
Formula Weight 307.2511
Percent Purity >95%
Chemical Name or Material 1',2'-Dehydro Dicyclomine
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.