missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)hydrazine Hyrochloride, TRC

Product Code. 30367172
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367172 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367172 Supplier Toronto Research Chemicals Supplier No. TRCC37552650G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Chlorophenyl)hydrazine Hyrochloride
CAS 41052-75-9
Molecular Formula C6H7N2Cl . (HCl)
Quantity 50 g
Synonym (2-Chlorophenyl)hydrazine Hyrochloride, (o-Chlorophenyl)hydrazine Hydrochloride, 1-(2-Chlorophenyl)hydrazine Hydrochloride, N-(2-Chlorophenyl)hydrazine Hydrochloride
InChI Formula InChI=1S/C6H7ClN2.ClH/c7-5-3-1-2-4-6(5)9-8;/h1-4,9H,8H2;1H
SMILES Cl.NNc1ccccc1Cl
IUPAC Name (2-chlorophenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 179.04
Formula Weight 178.0065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.