missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%, Thermo Scientific™

Product Code. p-7055759
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15488906

Brand: Thermo Scientific Alfa Aesar H64418.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406930-04-9
Molecular Formula C14H10ClF3N2O2
Molecular Weight (g/mol) 330.691
MDL Number MFCD02632826
InChI Key IBVFZYNARZAJSE-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea, 3-2-chlorophenyl-1-4-trifluoromethoxy phenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxyphenyl urea
PubChem CID 951288
IUPAC Name 1-(2-chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
SMILES C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 330.69
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.