missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Chloroethyl)pyrrolidine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Chloroethyl)pyrrolidine Hydrochloride |
| CAS | 7250-67-1 |
| Molecular Formula | C6H13Cl2N |
| Quantity | 25 g |
| Synonym | 1-(2-Chloroethyl)pyrrolidine Monohydrochloride, 1-(2-Chloroethyl)pyrrolidinium Chloride, 2-(1-Pyrrolidino)ethyl Chloride Hydrochloride, 2-(1-Pyrrolidinyl)ethyl Chloride Hydrochloride, 2-Pyrrolidinoethyl Chloride Hydrochloride, 2-Pyrrolidinyl-1-chloroethane Hydrochloride, N-(2-Chloroethyl)pyrrolidine Hydrochloride, N-(2-Chloroethyl)pyrrolidinium Chloride, N-(beta-Chloroethyl)pyrrolidine Hydrochloride, Pyrrolidinoethyl chloride hydrochloride, beta-Pyrrolidinoethyl Chloride Hydrochloride |
| InChI Formula | InChI=1S/C6H12ClN.ClH/c7-3-6-8-4-1-2-5-8;/h1-6H2;1H |
| SMILES | Cl.ClCCN1CCCC1 |
| IUPAC Name | 1-(2-chloroethyl)pyrrolidine;hydrochloride |
| Molecular Weight (g/mol) | 170.08 |
| Formula Weight | 169.0425 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?