missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)pyrrolidine Hydrochloride, TRC

Product Code. 30358423
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358423 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358423 Supplier Toronto Research Chemicals Supplier No. TRCC366235100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Chloroethyl)pyrrolidine Hydrochloride
CAS 7250-67-1
Molecular Formula C6H13Cl2N
Quantity 100 g
Synonym 1-(2-Chloroethyl)pyrrolidine Monohydrochloride, 1-(2-Chloroethyl)pyrrolidinium Chloride, 2-(1-Pyrrolidino)ethyl Chloride Hydrochloride, 2-(1-Pyrrolidinyl)ethyl Chloride Hydrochloride, 2-Pyrrolidinoethyl Chloride Hydrochloride, 2-Pyrrolidinyl-1-chloroethane Hydrochloride, N-(2-Chloroethyl)pyrrolidine Hydrochloride, N-(2-Chloroethyl)pyrrolidinium Chloride, N-(beta-Chloroethyl)pyrrolidine Hydrochloride, Pyrrolidinoethyl chloride hydrochloride, beta-Pyrrolidinoethyl Chloride Hydrochloride
InChI Formula InChI=1S/C6H12ClN.ClH/c7-3-6-8-4-1-2-5-8;/h1-6H2;1H
SMILES Cl.ClCCN1CCCC1
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
Molecular Weight (g/mol) 170.08
Formula Weight 169.0425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.