missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)pyrrolidine hydrochloride, 98%

Product Code. 11495876
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495876 25 g 25g
11405886 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495876 Supplier Thermo Scientific Alfa Aesar Supplier No. B22236.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7250-67-1 | C6H13Cl2N | 170.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7250-67-1
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012718
InChI Key FSNGFFWICFYWQC-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
PubChem CID 81668
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
SMILES C1CCN(C1)CCCl.Cl

Specifications

Melting Point 168°C to 173°C
Quantity 25 g
Beilstein 3677061
Sensitivity Hygroscopic
Formula Weight 170.08
Percent Purity 98%
Chemical Name or Material 1-(2-Chloroethyl)pyrrolidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.