Learn More
1-(2-Chloro-6-nitrophenyl)homopiperazine, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 874815-01-7 |
---|---|
Molecular Formula | C11H14ClN3O2 |
Molecular Weight (g/mol) | 255.702 |
InChI Key | DGSFYWNASDEFDR-UHFFFAOYSA-N |
Synonym | 1-2-chloro-6-nitrophenyl-1,4-diazepane, 1-6-chloro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-2-chloro-6-nitrophenyl hexahydro |
PubChem CID | 18526470 |
IUPAC Name | 1-(2-chloro-6-nitrophenyl)-1,4-diazepane |
SMILES | C1CNCCN(C1)C2=C(C=CC=C2Cl)[N+](=O)[O-] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
874815-01-7 | |
255.702 | |
1-2-chloro-6-nitrophenyl-1,4-diazepane, 1-6-chloro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-2-chloro-6-nitrophenyl hexahydro | |
1-(2-chloro-6-nitrophenyl)-1,4-diazepane |
C11H14ClN3O2 | |
DGSFYWNASDEFDR-UHFFFAOYSA-N | |
18526470 | |
C1CNCCN(C1)C2=C(C=CC=C2Cl)[N+](=O)[O-] |
Specifications
874815-01-7 | |
UN3259 | |
DGSFYWNASDEFDR-UHFFFAOYSA-N | |
1-(2-chloro-6-nitrophenyl)-1,4-diazepane | |
18526470 | |
96% | |
1-(2-Chloro-6-nitrophenyl)homopiperazine |
C11H14ClN3O2 | |
1-2-chloro-6-nitrophenyl-1,4-diazepane, 1-6-chloro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-2-chloro-6-nitrophenyl hexahydro | |
C1CNCCN(C1)C2=C(C=CC=C2Cl)[N+](=O)[O-] | |
255.702 | |
255.7 | |
Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures