missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Chloro-4-(4-chlorophenyl)butyl]-1H-imidazole (>85%), TRC

Product Code. 30354407
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354407 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354407 Supplier Toronto Research Chemicals Supplier No. TRCC378065250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 67085-12-5
Molecular Formula C13H14Cl2N2
Quantity 250 mg
Synonym Butoconazole Impurity 3
InChI Formula InChI=1S/C13H14Cl2N2/c14-12-4-1-11(2-5-12)3-6-13(15)9-17-8-7-16-10-17/h1-2,4-5,7-8,10,13H,3,6,9H2
SMILES ClC(CCc1ccc(Cl)cc1)Cn2ccnc2
IUPAC Name 1-[2-chloro-4-(4-chlorophenyl)butyl]imidazole
Molecular Weight (g/mol) 269.17
Formula Weight 268.0534
Chemical Name or Material 1-[2-Chloro-4-(4-chlorophenyl)butyl]-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.