missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromophenyl)piperazine, TRC

Product Code. 30347286
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347286 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347286 Supplier Toronto Research Chemicals Supplier No. TRCB80270510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Bromophenyl)piperazine
CAS 1011-13-8
Molecular Formula C10 H13 Br N2
Quantity 10 mg
InChI Formula InChI=1S/C10H13BrN2/c11-9-3-1-2-4-10(9)13-7-5-12-6-8-13/h1-4,12H,5-8H2
SMILES Brc1ccccc1N2CCNCC2
IUPAC Name 1-(2-bromophenyl)piperazine
Molecular Weight (g/mol) 241.128
Formula Weight 240.026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.