missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10451313
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 87488-84-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
MDL Number MFCD06659065
InChI Key QNDJHGODPWAKAO-UHFFFAOYSA-N
Synonym 1-2-bromophenyl-1h-pyrazole, 1-2-bromophenyl pyrazole, 1-2-bromo-phenyl-1h-pyrazole, 2-bromophenyl pyrazole, acmc-209qo1, 1h-pyrazole,1-2-bromophenyl, 1h-pyrazole, 1-2-bromophenyl
PubChem CID 2795433
IUPAC Name 1-(2-bromophenyl)-1H-pyrazole
SMILES BrC1=CC=CC=C1N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-(2-Bromophenyl)-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'