missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10406984
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10406984 5 g 5g
10451313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406984 Supplier Thermo Scientific Maybridge Supplier No. CC24410DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 87488-84-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
MDL Number MFCD06659065
InChI Key QNDJHGODPWAKAO-UHFFFAOYSA-N
Synonym 1-2-bromophenyl-1h-pyrazole, 1-2-bromophenyl pyrazole, 1-2-bromo-phenyl-1h-pyrazole, 2-bromophenyl pyrazole, acmc-209qo1, 1h-pyrazole,1-2-bromophenyl, 1h-pyrazole, 1-2-bromophenyl
PubChem CID 2795433
IUPAC Name 1-(2-bromophenyl)-1H-pyrazole
SMILES BrC1=CC=CC=C1N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-(2-Bromophenyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.