missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)-3-(trifluoromethyl)benzene, 95%, Thermo Scientific™

Product Code. 15411879
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1997-80-4
Molecular Formula C9H8BrF3
Molecular Weight (g/mol) 253.062
MDL Number MFCD03092900
InChI Key AZLYAPKPZBXCGX-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-3-trifluoromethyl benzene, 3-trifluoromethyl phenethyl bromide, 3-trifluoromethyl phenylethyl bromide, benzene, 1-2-bromoethyl-3-trifluoromethyl, 1-bromo-2-3-trifluoromethyl phenyl ethane, acmc-20anht, 3-trifluoromethylphenethyl bromide, toluene, m-2-bromoethyl-.alpha.,.alpha.,.alpha.-trifluoro, 3-trifluoromethyl phenylethylbromide
PubChem CID 594535
IUPAC Name 1-(2-bromoethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCBr

Specifications

Color Colorless
Density 1.495 g/mL
Boiling Point 92°C to 94°C (12 mmHg)
Refractive Index 1.486
Quantity 1 g
UN Number UN3082
Beilstein 1950052
Formula Weight 253.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'