missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-3-nitrobenzene, 97%, Thermo Scientific™

Catalog Number 10045982
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13831-59-9
C8H8BrNO3
246.06
MFCD07783648
QBUSKXLDUNPEMZ-UHFFFAOYSA-N
1-2-bromoethoxy-3-nitrobenzene, benzene,1-2-bromoethoxy-3-nitro, 2-bromoethyl 3-nitrophenyl ether, acmc-209chl, 2-bromo-3'-nitrophenetole, 2-3-nitrophenoxy ethyl bromide, 3-2-bromoethoxy-1-nitrobenzene
269626
1-(2-bromoethoxy)-3-nitrobenzene
[O-][N+](=O)C1=CC=CC(OCCBr)=C1
This item is not returnable. View return policy
13831-59-9
C8H8BrNO3
246.06
MFCD07783648
QBUSKXLDUNPEMZ-UHFFFAOYSA-N
1-2-bromoethoxy-3-nitrobenzene, benzene,1-2-bromoethoxy-3-nitro, 2-bromoethyl 3-nitrophenyl ether, acmc-209chl, 2-bromo-3'-nitrophenetole, 2-3-nitrophenoxy ethyl bromide, 3-2-bromoethoxy-1-nitrobenzene
269626
1-(2-bromoethoxy)-3-nitrobenzene
[O-][N+](=O)C1=CC=CC(OCCBr)=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 39°C
Color Brown or Green-Yellow
Boiling Point 140°C to 145°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H4OCH2CH2Br
Refractive Index 1.5830 to 1.585
Quantity 5 g
Formula Weight 246.06
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 1-(2-Bromoethoxy)-3-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Bromoethoxy)-3-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'