missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethylsilyloxy)ethane, 98%

Product Code. 10414345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10414345 50 g Glass bottle 50g
10700721 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414345 Supplier Thermo Scientific Acros Supplier No. 209580500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7381-30-8 | C8H22O2Si2 | 206.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7381-30-8
Molecular Formula C8H22O2Si2
Molecular Weight (g/mol) 206.43
MDL Number MFCD00009640
InChI Key JGWFUSVYECJQDT-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
PubChem CID 81858
IUPAC Name trimethyl(2-trimethylsilyloxyethoxy)silane
SMILES C[Si](C)(C)OCCO[Si](C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 165°C to 166°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
Refractive Index 1.4020 to 1.404
Quantity 50 g
Specific Gravity 0.84
Formula Weight 206.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(trimethylsilyloxy)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(trimethylsilyloxy)ethane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.