missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethoxysilyl)ethane, 96%

Product Code. 15467348
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467348 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467348 Supplier Thermo Scientific Alfa Aesar Supplier No. H60850.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18406-41-2 | C8H22O6Si2 | 270.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18406-41-2
Molecular Formula C8H22O6Si2
Molecular Weight (g/mol) 270.43
MDL Number MFCD00053672
InChI Key JCGDCINCKDQXDX-UHFFFAOYSA-N
Synonym 1,2-bis trimethoxysilyl ethane, 3,3,6,6-tetramethoxy-2,7-dioxa-3,6-disilaoctane, hexamethoxydisilylethane, 1,2-ethylenebis trimethoxysilane, dow corning x1-6145a, 2,7-dioxa-3,6-disilaoctane, 3,3,6,6-tetramethoxy, trimethoxy 2-trimethoxysilylethyl silane, 1,4-disilabutane, 1,1,1,4,4,4-hexamethoxy, acmc-209ele, ethylenebis trimethoxysilane
PubChem CID 87627
SMILES CO[Si](CC[Si](OC)(OC)OC)(OC)OC

Specifications

Color Colorless
Density 1.073 g/mL
Boiling Point 103°C to 104°C
Flash Point 109°C (228°F)
Linear Formula [-CH2Si(OCH3)3]2
Refractive Index 1.409
Quantity 5 mL
UN Number UN3382
IUPAC Name trimethoxy(2-trimethoxysilylethyl)silane
Formula Weight 270.43
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethoxysilyl)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.