missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylthio)ethane, 98+%

Product Code. 11321487
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321487 10 g 10g
11331487 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321487 Supplier Thermo Scientific Alfa Aesar Supplier No. L02019.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-20-8 | C14H14S2 | 246.39 g/mol

1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-20-8
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.39
MDL Number MFCD00014085
InChI Key MHCVYAFXPIMYRD-UHFFFAOYSA-N
Synonym 1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
PubChem CID 69317
IUPAC Name 2-phenylsulfanylethylsulfanylbenzene
SMILES C(CSC1=CC=CC=C1)SC1=CC=CC=C1

Specifications

Melting Point 68°C to 70°C
Linear Formula C6H5SCH2CH2SC6H5
Quantity 10 g
Beilstein 1877719
Solubility Information Insoluble in water.
Formula Weight 246.39
Percent Purity ≥98%
Chemical Name or Material 1,2-Bis(phenylthio)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.