missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)ethane, 97+%

Product Code. 11410263
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410263 5 g 5g
11420263 25 g 25g
11430263 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410263 Supplier Thermo Scientific Alfa Aesar Supplier No. A11419.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1663-45-2 | C26H24P2 | 398.43 g/mol

1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.

Solubility
Soluble in acetone, methanol and alcoholic hydrochloric acid.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-45-2
Molecular Formula C26H24P2
Molecular Weight (g/mol) 398.43
MDL Number MFCD00003047
InChI Key QFMZQPDHXULLKC-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
PubChem CID 74267
ChEBI CHEBI:30669
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138°C to 143°C
Linear Formula (C6H5)2PCH2CH2P(C6H5)2
Quantity 5 g
Beilstein 761261
Sensitivity Air Sensitive
Solubility Information Soluble in acetone,methanol and alcoholic hydrochloric acid.
Formula Weight 398.42
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis (diphenylphosphino)ethane
compliance-icons
Product Identifier
  • 1,2-Bis(diphenylphosphino)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Bis(diphenylphosphino)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.