missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylphosphino)ethane, 97%

Product Code. 11450147
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450147 0.25 g 0.25g
11440147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450147 Supplier Thermo Scientific Alfa Aesar Supplier No. B21051.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23936-60-9 | C6H16P2 | 150.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23936-60-9
Molecular Formula C6H16P2
Molecular Weight (g/mol) 150.14
MDL Number MFCD00008511
InChI Key ZKWQSBFSGZJNFP-UHFFFAOYSA-N
Synonym 1,2-bis dimethylphosphino ethane, ethylenebis dimethylphosphine, phosphine, 1,2-ethanediylbis dimethyl, 2-dimethylphosphanylethyl dimethyl phosphane, dimethyl-2-dimethylphosphino ethylphosphine, 2-dimethylphosphanyl ethyl dimethylphosphane, pubchem6482, acmc-1cjp0, ghl.pd_mitscher_leg0.173, zkwqsbfsgzjnfp-uhfffaoysa
PubChem CID 141059
IUPAC Name 2-dimethylphosphanylethyl(dimethyl)phosphane
SMILES CP(C)CCP(C)C

Specifications

Melting Point 179°C to 180°C
Density 0.9
Boiling Point 180°C
Flash Point −17°C (1.4°F)
Odor Recognizable
Linear Formula (CH3)2PCH2CH2P(CH3)2
Refractive Index 1.507
Quantity 0.25 g
UN Number UN1993
Sensitivity Air Sensitive
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 1,2-Bis(dimethylphosphino)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.