Learn More
1,2-Bis(dimethoxyphosphoryl)benzene, 99%, Thermo Scientific Chemicals
POA - £274.00
Chemical Identifiers
CAS | 15104-46-8 |
---|---|
Molecular Formula | C10H16O6P2 |
Molecular Weight (g/mol) | 294.18 |
MDL Number | MFCD00142461 |
InChI Key | TUKTVDDATWNXSN-UHFFFAOYSA-N |
Synonym | 1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one |
PubChem CID | 2733153 |
IUPAC Name | 1,2-bis(dimethoxyphosphoryl)benzene |
SMILES | COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363678
|
Thermo Scientific Alfa Aesar
039273.02 |
0.5 g | |||||||
11353678
|
Thermo Scientific Alfa Aesar
039273.04 |
2 g |
£68.60
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343678
|
Thermo Scientific Alfa Aesar
039273.09 |
10 g |
£274.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15104-46-8 | |
294.18 | |
TUKTVDDATWNXSN-UHFFFAOYSA-N | |
2733153 | |
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC |
C10H16O6P2 | |
MFCD00142461 | |
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one | |
1,2-bis(dimethoxyphosphoryl)benzene |
Specifications
15104-46-8 | |
C10H16O6P2 | |
UN3464 | |
TUKTVDDATWNXSN-UHFFFAOYSA-N | |
1,2-bis(dimethoxyphosphoryl)benzene | |
2733153 | |
99% | |
1,2-Bis(dimethoxyphosphoryl)benzene |
80°C to 82°C | |
MFCD00142461 | |
1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one | |
COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC | |
294.18 | |
294.18 | |
Crystalline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only