missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethoxyphosphoryl)benzene, 99%

Product Code. 11343678
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343678 10 g 10g
11363678 0.5 g 0.5g
11353678 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343678 Supplier Thermo Scientific Alfa Aesar Supplier No. 039273.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15104-46-8 | C10H16O6P2 | 294.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15104-46-8
Molecular Formula C10H16O6P2
Molecular Weight (g/mol) 294.18
MDL Number MFCD00142461
InChI Key TUKTVDDATWNXSN-UHFFFAOYSA-N
Synonym 1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one
PubChem CID 2733153
IUPAC Name 1,2-bis(dimethoxyphosphoryl)benzene
SMILES COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC

Specifications

Melting Point 80°C to 82°C
Quantity 10 g
UN Number UN3464
Formula Weight 294.18
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material 1,2-Bis(dimethoxyphosphoryl)benzene
compliance-icons
Product Identifier
  • 1,2-Bis(dimethoxyphosphoryl)benzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Bis(dimethoxyphosphoryl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.