missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diethylphosphino)ethane, 98%

Product Code. 15471527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471527 5 g 5g
15461527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471527 Supplier Thermo Scientific Alfa Aesar Supplier No. 039272.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6411-21-8 | C10H24P2 | 206.25 g/mol

It is used in chemical reaearch, organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6411-21-8
Molecular Formula C10H24P2
Molecular Weight (g/mol) 206.25
MDL Number MFCD00015175
InChI Key MIOCUERTSIJEDP-UHFFFAOYSA-N
Synonym 1,2-bis diethylphosphino ethane, bis 1,2-diethylphosphino ethane, 1,2-bis-diethylphosphino-ethane, acmc-1bbse, ethylenebis diethylphosphine, chembl70363, 1,2 bis diethylphosphino ethane, 1,2-bis-diethylphosphino ethane
PubChem CID 138745
SMILES CCP(CC)CCP(CC)CC

Specifications

Color Colorless
Density 0.884 g/mL
Boiling Point 124°C to 126°C (10 mmHg)
Odor Pungent
Linear Formula (CH3CH2)2PCH2CH2P(CH2CH3)2
Quantity 5 g
UN Number UN2845
Sensitivity Air Sensitive
Solubility Information It is soluble in organic solvents.
IUPAC Name 2-diethylphosphanylethyl(diethyl)phosphane
Formula Weight 206.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(diethylphosphino)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.