missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dichlorophosphino)ethane, 96%

Product Code. 11404557
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404557 5 g 5g
11494547 1 g 1g
11414557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404557 Supplier Thermo Scientific Alfa Aesar Supplier No. B21092.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 28240-69-9 | C2H4Cl4P2 | 231.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28240-69-9
Molecular Formula C2H4Cl4P2
Molecular Weight (g/mol) 231.802
MDL Number MFCD00009609
InChI Key SBWAJHLQMFBNIN-UHFFFAOYSA-N
PubChem CID 119904
IUPAC Name dichloro(2-dichlorophosphanylethyl)phosphane
SMILES C(CP(Cl)Cl)P(Cl)Cl

Specifications

Density 1.54
Boiling Point 70°C (1 mmHg)
Flash Point None
Refractive Index 1.588
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
Formula Weight 231.81
Percent Purity 96%
Chemical Name or Material 1,2-Bis(dichlorophosphino)ethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Bis(dichlorophosphino)ethane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Bis(dichlorophos

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.