missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2-Bis(carboxymethylthio)ethane, 97%

Product Code. 11424527
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11424527

Brand: Alfa Aesar™ B21074.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7244-02-2

Specifications

Melting Point 110°C
Molecular Formula C6H8O4S2
MDL Number MFCD00021771
Quantity 25 g
Synonym ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid
InChI Key CCQMORVULNZXIA-UHFFFAOYSA-L
SMILES C(CSCC(=O)O)SCC(=O)O
IUPAC Name 2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid
Molecular Weight (g/mol) 208.25
PubChem CID 310815
Formula Weight 210.26
Percent Purity 97%
Chemical Name or Material 1,2-Bis(carboxymethylthio)ethane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.