missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4-pyridyl)ethane, 97%

Product Code. 10702325 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702325 10 g Glass bottle 10g
10000600 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702325 Supplier Thermo Scientific Acros Supplier No. 106470100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4916-57-8 | C12H12N2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4916-57-8
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00006451
InChI Key DQRKTVIJNCVZAX-UHFFFAOYSA-N
Synonym 1,2-bis 4-pyridyl ethane, 1,2-di pyridin-4-yl ethane, 4,4'-ethylenedipyridine, 1,2-di 4-pyridyl ethane, 4,4'-ethane-1,2-diyldipyridine, unii-wo1evc302b, pyridine, 4,4'-1,2-ethanediyl bis, wo1evc302b, chembl70083, 4-2-pyridin-4-ylethyl pyridine
PubChem CID 78630
IUPAC Name 4-(2-pyridin-4-ylethyl)pyridine
SMILES C1=CN=CC=C1CCC2=CC=NC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 107.0°C to 113.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H4N)CH2CH2(C5H4N)
Quantity 10 g
Formula Weight 184.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Bis(4-pyridyl)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(4-pyridyl)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.