Learn More
1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, ACROS Organics™
Brand: Acros Organics 435212500
Code : 29
Additional Details : CAS Number : 1000393-36-1 Weight : 0.05025kg
Quantity | 250mg |
---|
Chemical Identifiers
1,2-Bis((2S,5S)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate | |
96.0 | |
97% | |
716.48 |
1000393-36-1 | |
100.0 | |
C34H56P2Rh·BF4 | |
MFCD07369040 |
Specifications
1,2-Bis((2S,5S)-2,5-Diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate | |
96.0 | |
54.7 to 59.4% C, 7.6 to 8.2% H | |
(1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure | |
97% | |
C34H56P2Rh·BF4 | |
MFCD07369040 | |
250mg |
1000393-36-1 | |
100.0 | |
Authentic | |
Powder | |
97% | |
716.48 | |
Yellow to orange to red to brown |