missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene, 97+%

Product Code. 15455576
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455576 250 mg 250mg
15445576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455576 Supplier Thermo Scientific Alfa Aesar Supplier No. H60227.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136705-65-2 | C26H44P2 | 418.586 g/mol

It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136705-65-2
Molecular Formula C26H44P2
Molecular Weight (g/mol) 418.586
MDL Number MFCD02684549
InChI Key RBVGOQHQBUPSGX-MOUTVQLLSA-N
Synonym r,r-i-pr-duphos, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, 1,2-bis 2r,5r-2,5-diisopropylphospholan-1-yl benzene, +-1,2-bis 2r,5r-2,5-di-i-propylphospholano benzene, +-1,2-bis 2r,5r-2,5-diisopropylphospholano benzene, kanata purity, 2r,5r-1-2-2r,5r-2,5-diisopropylphospholan-1-yl phenyl-2,5-diisopropylphospholane
PubChem CID 11327562
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-di(propan-2-yl)phospholan-1-yl]phenyl]-2,5-di(propan-2-yl)phospholane
SMILES CC(C)C1CCC(P1C2=CC=CC=C2P3C(CCC3C(C)C)C(C)C)C(C)C

Specifications

Flash Point >110°C (230°F)
Refractive Index 1.57
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Optical Rotation +103° (c=1 in Chloroform)
Formula Weight 418.58
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.