missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized

Product Code. 10570574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10570574 5 g Glass bottle 5g
10249673 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10570574 Supplier Thermo Scientific Acros Supplier No. 391690050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36839-55-1 | C6H12I2O2 | 369.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36839-55-1
Molecular Formula C6H12I2O2
Molecular Weight (g/mol) 369.96
MDL Number MFCD00075008
InChI Key BCAGFJXMCZSAHD-UHFFFAOYSA-N
Synonym 1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
PubChem CID 593436
IUPAC Name 1,2-bis(2-iodoethoxy)ethane
SMILES C(COCCI)OCCI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange-Red
Density 2.0280g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula I(CH2)2O(CH2)2O(CH2)2I
Quantity 5 g
Specific Gravity 2.028
Stabilizer copper
Formula Weight 369.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(2-iodoethoxy)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(2-iodoethoxy)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.