missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-chloroethoxy)ethane, 97%

Product Code. 11439937
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439937 100 g 100g
11449937 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439937 Supplier Thermo Scientific Alfa Aesar Supplier No. B22884.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-26-5 | C6H12Cl2O2 | 187.06 g/mol

1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-26-5
Molecular Formula C6H12Cl2O2
Molecular Weight (g/mol) 187.06
MDL Number MFCD00000976
InChI Key AGYUOJIYYGGHKV-UHFFFAOYSA-N
Synonym 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j
PubChem CID 8171
IUPAC Name 1,2-bis(2-chloroethoxy)ethane
SMILES ClCCOCCOCCCl

Specifications

Melting Point -32°C
Density 1.197
Boiling Point 235°C
Flash Point >110°C (230°F)
Refractive Index 1.461
Quantity 100 g
UN Number UN2810
Beilstein 506177
Solubility Information Insoluble in water.
Formula Weight 187.07
Percent Purity 97%
Chemical Name or Material 1,2-Bis(2-chloroethoxy)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(2-chloroethoxy)ethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Bis(2-chloroethoxy)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.