missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%

Product Code. p-67656 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10065670

Brand: Thermo Scientific Acros 401650010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 336624-09-0 | C22H20N2Na4O10 | 564.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336624-09-0
Molecular Formula C22H20N2Na4O10
Molecular Weight (g/mol) 564.37
MDL Number MFCD02683087
InChI Key ZWSMLJACYSGFKD-UHFFFAOYSA-J
Synonym BAPTA tetrasodium salt
IUPAC Name tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=C(OCCOC2=C(C=CC=C2)N(CC([O-])=O)CC([O-])=O)C=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Complexometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 564.36
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)-ethane-N,N,N′,N′-tetraacetic acid, tetrasodium salt hydrate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.