missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisoxazole-3-methanesulfonate Sodium Salt, TRC

Product Code. 30138265
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138265 1 g 1g
30132874 10 g 10g
30010756 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138265 Supplier Toronto Research Chemicals Supplier No. TRCB2019501G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73101-64-1
Molecular Formula C8 H6 N O4 S . Na
Molecular Weight (g/mol) 235.19
Synonym 1,2-Benzisoxazole-3-methanesulfonic acid, sodium salt (1:1), 1,2-Benzisoxazole-3-methanesulfonic acid sodium salt, 1,2-Benzisoxazole-3-methanesulfonic acid, sodium salt (9CI), Sodium 1,2-benzisoxazole-3-methanesulfonate
IUPAC Name sodium;1,2-benzoxazol-3-ylmethanesulfonate
SMILES [Na+].[O-]S(=O)(=O)Cc1noc2ccccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H7NO4S.Na/c10-14(11,12)5-7-6-3-1-2-4-8(6)13-9-7;/h1-4H,5H2,(H,10,11,12);/q;+1/p-1
Formula Weight 234.9915
Chemical Name or Material 1,2-Benzisoxazole-3-methanesulfonate, Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.