missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedithiol, 97%

Product Code. 11317656
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317656 250 mg 250mg
11397646 1 g 1g
11307656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11317656 Supplier Thermo Scientific Alfa Aesar Supplier No. L11253.MD
  • ONLINE EXCLUSIVE PRICE
    £42.00
    £37.65 / 250mg
    Save £4.35
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17534-15-5 | C6H6S2 | 142.23 g/mol

1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.

Notes
Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17534-15-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004835
InChI Key JRNVQLOKVMWBFR-UHFFFAOYSA-N
Synonym 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
PubChem CID 69370
IUPAC Name benzene-1,2-dithiol
SMILES SC1=C(S)C=CC=C1

Specifications

Melting Point 21°C to 25°C
Density 1.236
Boiling Point 239°C
Flash Point 103°C (217°F)
Odor Unpleasant
Refractive Index 1.658
Quantity 250 mg
UN Number UN3335
Beilstein 636154
Sensitivity Air sensitive
Solubility Information Soluble in water,benzene,ethanol,diethyl ether,ethyl acetate.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedithiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.