missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedimethanol, 97%

Product Code. 11456176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456176 1 g 1g
11466176 5 g 5g
11476176 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456176 Supplier Thermo Scientific Alfa Aesar Supplier No. B23008.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

1,2-Benzenedimethanol is a raw material used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.

Solubility
Soluble in water, ether, ethanol, benzene.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

Melting Point 59°C to 64°C
Quantity 1 g
Beilstein 508775
Solubility Information Soluble in water,ether,ethanol,benzene.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedimethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.