missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedimethanol, 97%

Product Code. 10521564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
This item is not returnable. View return policy

Product Code. 10521564

Brand: Thermo Scientific Acros 180410100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White to Yellow
Density 145g/mL
Boiling Point 291.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CH2OH)2
Quantity 10 g
Beilstein 06, 910
Fieser 16,18; 12,563; 13,341
Solubility Information Solubility in water: soluble
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 2-Benzenedimethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.