missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedicarboxylic Acid, Dipentylester, Branched and Linear, TRC

Product Code. 30179008
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179008 25 mg 25mg
30179431 5 mg 5mg
30179263 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179008 Supplier Toronto Research Chemicals Supplier No. TRCB18931025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84777-06-0
Molecular Formula C18 H26 O4
Molecular Weight (g/mol) 306.4
Synonym 1,2-Benzenedicarboxylic acid, dipentyl ester, branched and linear, Dipentyl phthalate branched and linear, Phthalic acid, n-pentyl-isopentyl ester (mixture of isomers)
IUPAC Name dipentan-2-yl benzene-1,2-dicarboxylate
SMILES CCCC(C)OC(=O)c1ccccc1C(=O)OC(C)CCC

Specifications

Analyte or Component Names Dipentyl phthalate (mix of isomers)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C18H26O4/c1-5-9-13(3)21-17(19)15-11-7-8-12-16(15)18(20)22-14(4)10-6-2/h7-8,11-14H,5-6,9-10H2,1-4H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 mg
Formula Weight 306.18
Percent Purity >95%
Chemical Name or Material Dipentyl phthalate (mix of isomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.