missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzenedicarboxylic Acid 1-(7-Methyloctyl) Ester, TRC
Specifications
Specifications
| Analyte or Component Names | 1,2-Benzenedicarboxylic Acid 1-(7-Methyloctyl) Ester |
| Shipping Condition | Room Temperature |
| InChI Formula | InChI=1S/C17H24O4/c1-13(2)9-5-3-4-8-12-21-17(20)15-11-7-6-10-14(15)16(18)19/h6-7,10-11,13H,3-5,8-9,12H2,1-2H3,(H,18,19) |
| Solution Type | Neat |
| Industry Type | Pharmaceutical |
| Quantity | 500 mg |
| Formula Weight | 292.1675 |
| Chemical Name or Material | 1,2-Benzenedicarboxylic Acid 1-(7-Methyloctyl) Ester |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?