missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperazine, 99%

Product Code. p-54599 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249400 100 g Glass bottle 100g
10259400 500 g Glass bottle 500g
10258753 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249400 Supplier Thermo Scientific Acros Supplier No. 400761000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-31-8 | C6H15N3 | 129.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 140-31-8
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.21
MDL Number MFCD00005971
InChI Key IMUDHTPIFIBORV-UHFFFAOYSA-N
Synonym n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
PubChem CID 8795
IUPAC Name 2-piperazin-1-ylethanamine
SMILES NCCN1CCNCC1

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -19.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 218.0°C to 222.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5010
Quantity 100 g
Specific Gravity 0.98
Solubility Information Solubility in water: soluble.
Viscosity 14 mPa.s (20°C)
Formula Weight 129.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-(2-Aminoethyl)piperazine, 99%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.