missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Aminoethyl)piperazine, 98%

Brand:  Alfa Aesar™ A10154.0B

Code : NEW

Additional Details : CAS Number : 140-31-8 Weight : 1.00000kg Transport : UN number : 2815 Chem class : 1b Pack group : III

Quantity 1000g

 View more versions of this product

Product Code. 11499422

  • £37.70 / 1000g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Description

Description

Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
Specifications

Chemical Identifiers

1- (2-Aminoethyl)piperazine
C6H15N3
MFCD00005971
104363
140-31-8
129.21
UN2815
1-Piperazineethylamine; 2-(1-Piperazinyl)ethylamine

Specifications

1- (2-Aminoethyl)piperazine
−19°C
98%
93°C (199°F)
129.21
UN2815
1-Piperazineethylamine; 2-(1-Piperazinyl)ethylamine
140-31-8
222°C
1000g
C6H15N3
MFCD00005971
104363
Videos
SDS