missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
13.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1000 g
Description
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
Specifications
Melting Point | -19°C |
Density | 0.983 |
Boiling Point | 222°C |
Flash Point | 93°C (199°F) |
Refractive Index | 1.5 |
Quantity | 250 g |
UN Number | UN2815 |
Beilstein | 104363 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'