missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol, 96%

Product Code. 11352336
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352336 25 g 25g
11362336 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352336 Supplier Thermo Scientific Alfa Aesar Supplier No. L06032.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-69-4 | C6H14O3 | 134.18 g/mol

1,2,6-Hexanetriol is used as moisturizing agent (cremes), as solvent (pharmaceuticals) and as crosslinker in plastics. It acts as wetting agent of tobacco and special hydraulic fluid. It is used in the production of alkyd resins, celluloid, polyurethane and synthetic rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,6-Hexanetriol is used as moisturizing agent (cremes), as solvent (pharmaceuticals) and as crosslinker in plastics. It is used as rocket propellant. It acts as wetting agent of tobacco and special hydraulic fluid. It is used in the production of alkyd resins, celluloid, polyurethane and synthetic rubber.

Solubility
Miscible in water and acetone. Soluble in methanol.

Notes
Hygroscopic. Incompatible with Strong oxidizing agents, Acid chlorides, Acid anhydrides. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-69-4
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002976
InChI Key ZWVMLYRJXORSEP-UHFFFAOYNA-N
Synonym 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
PubChem CID 7823
IUPAC Name hexane-1,2,6-triol
SMILES OCCCCC(O)CO

Specifications

Melting Point -20°C
Density 1.106
Boiling Point 166°C to 168°C (2 mmHg)
Flash Point 191°C (375°F)
Refractive Index 1.476
Quantity 25 g
Beilstein 1304479
Sensitivity Hygroscopic
Solubility Information Miscible in water and acetone. Soluble in methanol.
Formula Weight 134.18
Percent Purity 96%
Chemical Name or Material 1,2,6-Hexanetriol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.