missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Dimethylphenoxy)acetone, TRC

Product Code. 30135562
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135562 10 g 10g
17938424 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135562 Supplier Toronto Research Chemicals Supplier No. TRCD47644010G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53012-41-2
Molecular Formula C11 H14 O2
Molecular Weight (g/mol) 178.23
Synonym 1-(2,6-Dimethylphenoxy)propan-2-one, Mexiletine Hydrochloride Imp. B (EP), 2-Propanone, 1-(2,6-dimethylphenoxy)-, 1-(2,6-Dimethylphenoxy)-2-propanone, 1-(2,6-Dimethylphenoxy)acetone
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-one
SMILES CC(=O)COc1c(C)cccc1C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H14O2/c1-8-5-4-6-9(2)11(8)13-7-10(3)12/h4-6H,7H2,1-3H3
Formula Weight 178.0994
Percent Purity >95%
Chemical Name or Material 1-(2,6-Dimethylphenoxy)propan-2-one
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.