missing translation for 'onlineSavingsMsg'
Learn More

1,2,5-Oxadiazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. p-3110043
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10259292

Brand: Thermo Scientific Maybridge CC07901CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 88598-08-7
Molecular Formula C3H2N2O3
Molecular Weight (g/mol) 114.06
MDL Number MFCD00604386
InChI Key JBLHCUQCDKBPGY-UHFFFAOYSA-N
Synonym furazan-3-carboxylic acid, 3-carboxy-1,2,5-oxadiazole, 1,2,5-oxadiazole-3-carboxylicacid, acmc-1bkac, 1,2,5-oxadiazole, oxd4, 9ci
PubChem CID 1415524
IUPAC Name 1,2,5-oxadiazole-3-carboxylic acid
SMILES OC(=O)C1=NON=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 114.06
Percent Purity 97%
Chemical Name or Material 1, 2, 5-Oxadiazole-3-carboxylic acid
compliance-icons
Product Identifier
  • 1,2,5-Oxadiazole-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,2,5-Oxadiazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.