missing translation for 'onlineSavingsMsg'
Learn More

1-(2’,5’-Dimethoxyphenyl)-2-chloroethanone, TRC

Product Code. 30382483
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382483 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382483 Supplier Toronto Research Chemicals Supplier No. TRCD470725100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2'­,5'­Dimethoxyphenyl)-2-chloroethanone
CAS 1204-22-4
Molecular Formula C10 H11 Cl O3
Quantity 100 mg
InChI Formula InChI=1S/C10H11ClO3/c1-13-7-3-4-10(14-2)8(5-7)9(12)6-11/h3-5H,6H2,1-2H3
SMILES COc1ccc(OC)c(c1)C(=O)CCl
IUPAC Name 2-chloro-1-(2,5-dimethoxyphenyl)ethanone
Molecular Weight (g/mol) 214.65
Formula Weight 214.0397
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.