missing translation for 'onlineSavingsMsg'
Learn More

1,2,5,6,9,10-Hexabromocyclododecane, TRC

Product Code. 30389419
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389419 25 g 25g
30396172 10 g 10g
30393575 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389419 Supplier Toronto Research Chemicals Supplier No. TRCH29074025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3194-55-6
Molecular Formula C12 H18 Br6
Molecular Weight (g/mol) 641.7
Synonym Cyclododecane, 1,2,5,6,9,10-hexabromo-, 1,2,5,6,9,10-Hexabromocyclododecane, Bromkal 73-6D, FR 1206, FR 1206HT, Pyroguard SR 104, SR 104, YM 88A, Hexabromocyclododecane, HBCD, HBCDD
IUPAC Name 1,2,5,6,9,10-hexabromocyclododecane
SMILES BrC1CCC(Br)C(Br)CCC(Br)C(Br)CCC1Br

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2
Formula Weight 635.6509
Chemical Name or Material 1,2,5,6,9,10-Hexabromocyclododecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.