missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethylbenzene, 98%

Product Code. 10319043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10319043 10 L Plastic drum 10 L
10786151 1 L Glass Bottle 1 L
10486960 2.5 L Glass Bottle 2.5 L
15139183 25 L Plastic drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10319043 Supplier Thermo Scientific Acros Supplier No. 140090100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-63-6 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-63-6
MDL Number MFCD00008527
InChI Key GWHJZXXIDMPWGX-UHFFFAOYSA-N
Synonym pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene
PubChem CID 7247
ChEBI CHEBI:34039
IUPAC Name 1,2,4-trimethylbenzene
SMILES CC1=CC(=C(C=C1)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -44.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 168.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Molecular Formula C9H12
Linear Formula C6H3(CH3)3
Refractive Index 1.5030 to 1.5050
Quantity 10 L
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and ether
Molecular Weight (g/mol) 120.195
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2,4-Trimethylbenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Trimethylbenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.