missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trifluorobenzene, 98+%

Product Code. 11410273
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410273 25 g 25g
11400273 5 g 5g
11420273 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11424.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-23-7 | C6H3F3 | 132.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-23-7
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.09
MDL Number MFCD00000305
InChI Key PEBWOGPSYUIOBP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trifluoro, unii-qww4hnq0l1, 1,3,4-trifluorobenzene, qww4hnq0l1, pubchem1068, 1,4-trifluorobenzene, benzene,2,4-trifluoro, 1,2,4-trifluoro-benzene, 1,2,4-trifluorobenzene, acmc-209io5
PubChem CID 67773
IUPAC Name 1,2,4-trifluorobenzene
SMILES FC1=CC=C(F)C(F)=C1

Specifications

Density 1.264
Boiling Point 89°C to 91°C
Flash Point 4°C (39°F)
Refractive Index 1.423
Quantity 25 g
UN Number UN2387
Beilstein 2042865
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,2,4-Trifluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.