missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%, Honeywell Riedel-de Haën™

Product Code. 15632770
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632770 1 L Glass Bottle 1 L
15642770 2 L Glass Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15632770 Supplier Honeywell-Chromasolv Supplier No. 2564121L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CHROMASOLV™,≥99%

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000547
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 16°C (literature)
CAS Max % ≤100.0000%
Density 1.454 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 105°C
Packaging Glass Bottle
Linear Formula C6H3Cl3
UN Number UN2321
Beilstein 956819
Boiling Point 213.5°C
Formula Weight 181.46
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1,2,4-Trichlorobenzene, CHROMASOLV™, ≥99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Trichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.